missing translation for 'onlineSavingsMsg'
Learn More

Silver cyanate, 98+%

Product Code. 10154610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10154610 25 g Glass bottle 25g
10012810 10 g Glass bottle 10g
10563391 50 g Glass bottle 50g
11954351 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10154610 Supplier Thermo Scientific Acros Supplier No. 223710250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3315-16-0 | CAgNO | 149.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3315-16-0
Molecular Formula CAgNO
Molecular Weight (g/mol) 149.89
InChI Key VAQVDZZAAMAZAO-UHFFFAOYSA-N
Synonym silver,cyanate, cyanic acid; silver
PubChem CID 45051745
IUPAC Name cyanic acid;silver
SMILES C(#N)O.[Ag]

Specifications

Boiling Point 1085.0°C
Melting Point 652.0°C
Color Beige to Gray
Physical Form Powder
Quantity 25 g
Assay Percent Range 98% min
Linear Formula AgOCN
Fieser 02,224; 03,162
Solubility Information Solubility in water: soluble
Formula Weight 149.89
Percent Purity ≥98%
Packaging Glass bottle
Chemical Name or Material Silver cyanate
Show More Show Less
compliance-icons
Product Identifier
  • Silver cyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.