missing translation for 'onlineSavingsMsg'
Learn More

Silver cyanate, 98+%, ACROS Organics™

Brand:  Acros Organics 223710100

Code : 29

Additional Details : CAS Number : 3315-16-0 Weight : 0.06000kg


Packaging Glass bottle
Quantity 10g

 View more versions of this product

Product Code. 10012810

  • £55.37 / 10g

Estimated Shipment
24-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

3315-16-0
149.89
silver,cyanate, cyanic acid; silver
cyanic acid;silver
CAgNO
VAQVDZZAAMAZAO-UHFFFAOYSA-N
45051745
C(#N)O.[Ag]

Specifications

Silver cyanate
98% min
AgOCN
silver,cyanate, cyanic acid; silver
VAQVDZZAAMAZAO-UHFFFAOYSA-N
cyanic acid;silver
45051745
Powder
Glass bottle
1085.0°C
652.0°C
3315-16-0
CAgNO
02,224; 03,162
Solubility in water: soluble
C(#N)O.[Ag]
149.89
149.89
98+%
Solubility in water: soluble.
Beige to Gray
10g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Silver cyanate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS Documents