missing translation for 'onlineSavingsMsg'
Learn More

Silver Chloride, 99%, ReagentPlus™, Honeywell™

Product Code. p-7120761 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
50g
This item is not returnable. View return policy

Product Code. 15672260

Brand: Honeywell Research Chemicals 22792710G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Boiling Point 1547°C
Melting Point 455°C
Color White
Quantity 10 g
UN Number UN3077
Formula Weight 143.32g/mol
Percent Purity 99%
Grade ReagentPlus™
Packaging Glass Bottle
Appearance Powder or Powder with Chunk(s)
Vapor Pressure 1mmHg (912°C)
Density 5.54g/cm3
Chemical Name or Material Silver Chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.