Learn More
Silver chloride, 95%
Description
Employed in Silver plating. Owing to its characteristic of reversible reduction to silver metal, it is used in photochromic lenses. Used as a cathode in sea water activated batteries. In electrochemistry, silver chloride electrode is used for potentiometric measurements. It serves as an antidote for mercury poisoning, and eliminates mercury from body. It is used in glass manufacturing industry. It is useful in the production of bandages, wound healing products and inglaze lustre, personal deodorant products, as well as for long term preservation of drinking water in water tanks; its pharmaceutical composition finds use as an antibacterial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1550°C |
| Melting Point | 455°C |
| Color | White |
| Physical Form | Coarse Powder |
| Quantity | 100 g |
| UN Number | UN3077 |
| Merck Index | 14,8509 |
| Solubility Information | Practically insoluble in water; Soluble in ammonia,solutions of alkali cyanides,thiosulfates and ammonium carbonates; Insoluble in alcohol and dilute acids. |
| Formula Weight | 143.32 |
| Percent Purity | 95% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.