missing translation for 'onlineSavingsMsg'
Learn More

Silver carbonate, 99%, pure, Thermo Scientific Chemicals

Product Code. 10694862
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-16-7
Molecular Formula CAg2O3
Molecular Weight (g/mol) 275.75
MDL Number MFCD00003403
InChI Key WIKQEUJFZPCFNJ-UHFFFAOYSA-N
Synonym carbonic acid; silver, carbonic acid disilver
PubChem CID 25203940
IUPAC Name carbonic acid;silver
SMILES C(=O)(O)O.[Ag].[Ag]

Specifications

Melting Point 210.0°C
Color Green to Green-Yellow
Physical Form Granular Powder
Quantity 100 g
Assay Percent Range 99%
Linear Formula Ag2CO3
Fieser 01,1005; 02,363; 04,425; 06,511; 08,411; 12,432
Merck Index 15, 8646
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonium hydroxide and nitric acid, insoluble in alcohol
Formula Weight 275.75
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Silver carbonate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'