missing translation for 'onlineSavingsMsg'
Learn More

Silver bromide, 99.5%

Product Code. 11381249
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381249 25 g 25g
11301259 5 g 5g
11391249 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381249 Supplier Thermo Scientific Alfa Aesar Supplier No. 011425.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7785-23-1 | AgBr | 187.772 g/mol

Silver bromide is used as the light-sensitive component on ordinary photographic film and plates and also used in I.R. applications and ionic semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-23-1
Molecular Formula AgBr
Molecular Weight (g/mol) 187.772
MDL Number MFCD00003398
InChI Key ADZWSOLPGZMUMY-UHFFFAOYSA-M
Synonym silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion
PubChem CID 66199
IUPAC Name bromosilver
SMILES Br[Ag]

Specifications

Melting Point 432°C
Color Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9%
Solubility Information Insoluble in water,alcohol and acids. Partially soluble in ammonia.
Formula Weight 187.78
Odor Odorless
Sensitivity Light sensitive
Appearance Pale yellow powder/flake/crystalline/beads
Density 6.47
Chemical Name or Material Silver bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.