Learn More
Silver, brazing flux, white paste, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 041355.TD
Code : NEW
Additional Details : CAS Number : 7440-22-4 Weight : 0.45400kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Silver, brazing flux | |
White | |
MFCD01868499 |
Active range: 540 to 980° | |
454 g |