missing translation for 'onlineSavingsMsg'
Learn More

Silver benzoate hydrate

Product Code. 11331998
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331998 25 g 25g
11341998 5 g 5g
11321998 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331998 Supplier Thermo Scientific Alfa Aesar Supplier No. 011896.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 532-31-0 | C7H5AgO2 | 228.98 g/mol

Silver benzoate hydrate along with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene (MTBD) is an efficient catalytic system for the reaction of carbon dioxide with various ketones. It was used in the synthesis of triphenyltinbenzoate. It is also used as a catalyst in Wolff rearrangement reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-31-0
Molecular Formula C7H5AgO2
Molecular Weight (g/mol) 228.98
MDL Number MFCD00013030
InChI Key CLDWGXZGFUNWKB-UHFFFAOYSA-M
Synonym silver benzoate hydrate
PubChem CID 87384783
SMILES [Ag+].[O-]C(=O)C1=CC=CC=C1

Specifications

Color Cream to White or Gray
Physical Form Powder
Quantity 25 g
Linear Formula C6H5CO2Ag·xH2O
Solubility Information Insoluble in water.
IUPAC Name benzoic acid silver
Formula Weight 228.89 (Anhydrous)
Sensitivity Light sensitive
Chemical Name or Material Silver benzoate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.