missing translation for 'onlineSavingsMsg'
Learn More

Silver benzoate hydrate

Product Code. 11321998
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11321998

Brand: Thermo Scientific Alfa Aesar 011896.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 532-31-0 | C7H5AgO2 | 228.98 g/mol

Silver benzoate hydrate along with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene (MTBD) is an efficient catalytic system for the reaction of carbon dioxide with various ketones. It was used in the synthesis of triphenyltinbenzoate. It is also used as a catalyst in Wolff rearrangement reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-31-0
Molecular Formula C7H5AgO2
Molecular Weight (g/mol) 228.98
MDL Number MFCD00013030
InChI Key CLDWGXZGFUNWKB-UHFFFAOYSA-M
Synonym silver benzoate hydrate
PubChem CID 87384783
SMILES [Ag+].[O-]C(=O)C1=CC=CC=C1

Specifications

Color Cream to White or Gray
Physical Form Powder
Quantity 100 g
Linear Formula C6H5CO2Ag·xH2O
Solubility Information Insoluble in water.
IUPAC Name benzoic acid;silver;hydrate
Formula Weight 228.89 (Anhydrous)
Sensitivity Light sensitive
Chemical Name or Material Silver benzoate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.