missing translation for 'onlineSavingsMsg'
Learn More

Versilon™ Silver Silver Antimicrobial Tubing

Product Code. 30053790 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/8 in.
Diameter (English) Outer:
1/2 in.
1/4 in.
3/4 in.
3/8 in.
5/16 in.
Unit Size:
15m
This item is not returnable. View return policy

Product Code. 30053790

Brand: Versilon™ Silver 5010718

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Silver-based antimicrobial compound inhibits growth of microbes

Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Plasticizer-free inner bore—excellent for food and beverage applications. Reduction in microbe growth effectively minimizes foul odors and discoloration. Meets FDA criteria. Phthalate and BPA free

Specifications

Certifications/Compliance FDA;Non-Phthalate;NSF-51
Diameter (English) Inner 1/2 in.
Diameter (English) Outer 3/4 in.
Overall Length (Metric) 50 ft.
Color Silver
Product Type Antimicrobial Tubing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.