missing translation for 'onlineSavingsMsg'
Learn More

Silver acetate, 99%, pure

Product Code. 15254674 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15254674 500 g 500g
10402401 25 g 25g
10542991 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15254674 Supplier Thermo Scientific Acros Supplier No. 202395000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 563-63-3 | C2H3AgO2 | 166.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-63-3
Molecular Formula C2H3AgO2
Molecular Weight (g/mol) 166.91
MDL Number MFCD00012446
InChI Key CQLFBEKRDQMJLZ-UHFFFAOYSA-M
Synonym silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
PubChem CID 11246
IUPAC Name silver;acetate
SMILES CC(=O)[O-].[Ag+]

Specifications

Color Beige-Gray to White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99%
Linear Formula CH3CO2Ag
Fieser 01,1002; 03,362; 06,511; 08,440; 12,256; 13,285
Merck Index 15,8644
Solubility Information Solubility in water: 10.2g/L (20°C). Other solubilities: soluble in nitric acid
Formula Weight 166.91
Percent Purity 99%
Grade Pure
Infrared Spectrum Authentic
Specific Gravity 3.25
Density 3.2500g/mL
Chemical Name or Material Silver acetate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.