missing translation for 'onlineSavingsMsg'
Learn More

Silver, 99.9%, (trace metal basis), wire, 0.5mm diam., length 10m, 1ea = 20gr

Product Code. 10232462 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10232462 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10232462 Supplier Thermo Scientific Acros Supplier No. 317710010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
CAS 7440-22-4
Melting Point 961.0°C
Boiling Point 2212.0°C
Quantity 1 Ea.
Assay Percent Range 99.90%
Form Wire
Molecular Formula Ag
Linear Formula Ag
MDL Number MFCD00003397
Fieser 04,164
Merck Index 15, 8643
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
SMILES [Ag]
IUPAC Name silver
Molecular Weight (g/mol) 107.87
PubChem CID 23954
ChEBI CHEBI:9141
Formula Weight 107.87
Percent Purity 99.9%
Grade Trace Metal Basis
Chemical Name or Material Silver, 99.9% (1ea = 20gr)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.