missing translation for 'onlineSavingsMsg'
Learn More

Silver 2,4-pentanedionate, 98%, Thermo Scientific Chemicals

Product Code. 15411288
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Silver 2,4-pentanedionate is used to make silver impregnated nanocrystalline metal oxides, with special emphasis on aluminum oxide. It is used as a cationization reagent in combination with polar matrices such as 2,5-dihydroxybenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silver 2,4-pentanedionate is used to make silver impregnated nanocrystalline metal oxides, with special emphasis on aluminum oxide. It is used as a cationization reagent in combination with polar matrices such as 2,5-dihydroxybenzoic acid.

Solubility
Soluble in water.

Notes
Air, light and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15525-64-1
Molecular Formula C5H8AgO2
Molecular Weight (g/mol) 207.985
MDL Number MFCD00000012
InChI Key LEUOXKLUFCTIIY-LNKPDPKZSA-N
Synonym silver acetylacetonate, 2,4-pentanedione, silver derivative, silver, 2,4-pentanedionato-o,o', silver 2,4-pentanedionate, 4-oxopent-2-en-2-yl oxy silver, 3z-4-argentiooxy pent-3-en-2-one
PubChem CID 11074572
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;silver
SMILES CC(=CC(=O)C)O.[Ag]

Specifications

Melting Point 100°C (decomposition)
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 206.98
Percent Purity 98%
Odor Characteristic
Sensitivity Air, light, and moisture sensitive
Chemical Name or Material Silver 2,4-pentanedionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'