missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Thermo Scientific Split / Splitless Tapered FAST FocusLiner

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 09211101

Code : 13

Additional Details : Weight : 0.00990kg

 View more versions of this product

Product Code. 12964041

  • £39.40 / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket

Description

Description

    Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Thermo Scientific instruments
Specifications

Specifications

GC Inlet liner, Tapered FocusLiner
78.5 mm
Finnigan 9001 and GCQ GCs, and Trace 1300, 1310
1 Ea.
2.3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers