missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Thermo Scientific Split / Splitless Tapered FAST FocusLiner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 092115
Code : 13
Additional Details : Weight : 0.80000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Thermo Scientific instruments
Specifications
GC Inlet liner, Tapered FocusLiner | |
78.5 mm | |
6.3 mm |
25/Pk. | |
2.3 mm | |
Finnigan 9001 and GCQ GCs, and Trace 1300, 1310 |