Learn More
Trajan Scientific and Medical™ SilTite™ Ferrules
Ensures easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 073228
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15218923
  • £84.30 / Pack of 10
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
For Use with Shimadzu Detector/Injectors, Standard MS Interface 0.32 mm I.D. Columns
SilTite Ferrules for 0.32 mm ID Columns for Shimadzu