Learn More
Trajan Scientific and Medical™ SilTite™ Column Connectors
Ensures easy, leak-free installation for capillary columns without the use of any tools.
Brand: Trajan Scientific and Medical™ 073219
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
SilTite Kit 1/32 in. Column for PerkinElmer | |
1/32 in. | |
Column Connectors | |
Installation for Capillary Columns |