Learn More
Trajan Scientific and Medical™ SilTite™ Column Connectors
Ensures easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 073273
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15258923
  • £94.30 / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
SilTite Kit 1/32 in. Column for Agilent
1/32 in. column size
Installation for Capillary Columns