missing translation for 'onlineSavingsMsg'
Learn More

Siloxane Mixture 61 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30317409
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317409 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317409 Supplier Dr. Ehrenstorfer Supplier No. DREA30000061AL
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclomethicone 5∣Decamethyltetrasiloxane∣Dodecamethylcyclohexasiloxane∣Dodecamethylpentasiloxane∣Hexamethylcyclotrisiloxane∣Hexamethyldisiloxane∣Octamethylcyclotetrasiloxane∣Octamethyltrisiloxane∣Tetradecamethylhexasiloxane
CAS 541-02-6, 141-62-8, 540-97-6, 141-63-9, 541-05-9, 107-46-0
Analyte or Component Names Cyclomethicone 5∣Decamethyltetrasiloxane∣Dodecamethylcyclohexasiloxane∣Dodecamethylpentasiloxane∣Hexamethylcyclotrisiloxane∣Hexamethyldisiloxane∣Octamethylcyclotetrasiloxane∣Octamethyltrisiloxane∣Tetradecamethylhexasiloxane
Shipping Condition +20°C
Certifications/Compliance ISO/IEC 17025
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Cyclomethicone 5 100 μg/mL∣Decamethyltetrasiloxane 100 μg/mL∣Dodecamethylcyclohexasiloxane 100 μg/mL∣Dodecamethylpentasiloxane 100 μg/mL∣Hexamethylcyclotrisiloxane 100 μg/mL∣Hexamethyldisiloxane 100 μg/mL∣Octamethylcyclotetrasiloxane 100 μg/mL∣Octamethyltrisiloxane 100 μg/mL∣Tetradecamethylhexasiloxane 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.