missing translation for 'onlineSavingsMsg'
Learn More

Silodosin, MedChemExpress

Product Code. p-200097148
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16434556

Brand: Medchem Express HY10122

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Silodosin (KAD 3213; KMD 3213) is a potent, selective and orally active α1A-adrenergic receptor (α1A-AR) blocker. Silodosin exhibits high affinity for α1A-AR (Ki=0.036 nM), over 162-fold and 50-fold than for α1B-AR and α1D-AR with Ki values of 21 nM and 2.0 nM, respectively. Silodosin is an effective and well-tolerated agent, it can be used for the investigation of LUTS/BPH.

Silodosin (KAD 3213

Specifications

Chemical Name or Material Silodosin
CAS 160970-54-7
Color White
Purity Grade Notes Research
Molecular Formula C25H32F3N3O4
Quantity 100 mg
Synonym KAD 3213 KMD 3213
Solubility Information DMSO : ≥ 50 mg/mL (100.90 mM)
SMILES O=C(N)C1=CC(C[C@@H](C)NCCOC2=CC=CC=C2OCC(F)(F)F)=CC3=C1N(CC3)CCCO
Molecular Weight (g/mol) 495.53
Formula Weight 495.53
Percent Purity 95.07%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.