missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Silodosin
GREENER_CHOICE

Product Code. 18797376
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18797376

Brand: Tocris Bioscience™ 6663/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective α1A adrenoceptor antagonist (Ki = 0.036 nM)

Silodosin is a selective α1A adrenoceptor antagonist (Ki = 0.036 nM). Displays >55 fold selectivity for α1A over α1B and α1D. Inhibits norepinephrine-induced [Ca2+]I increase in α1A expressing CHO cells (IC50 = 0.32 nM). Inhibits norepinephrine-induced contraction of isolated human prostate tissue.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dihydro-1-(3-hydroxypropyl)-5-[(2R)-2-[[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl]amino]propyl]-1H-indole-7-carboxamide
CAS 160970-54-7
Quantity 10 mg
Molecular Formula C25H32F3N3O4
Formula Weight 495.53
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.