missing translation for 'onlineSavingsMsg'
Learn More

Silmitasertib sodium salt, MedChemExpress

Product Code. 16490568
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490568 5 mg 5mg
16408118 10 mg 10mg
16484027 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490568 Supplier Medchem Express Supplier No. HY50855B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Silmitasertib sodium salt is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.

Silmitasertib sodium salt is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.

Specifications

Chemical Name or Material Silmitasertib sodium salt
CAS 1309357-15-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H11ClN3NaO2
Quantity 5 mg
Synonym CX-4945 sodium salt
Solubility Information H2O : 16.67 mg/mL (44.84 mM; Need ultrasonic) ∣DMSO : 6.67 mg/mL (17.94 mM; Need ultrasonic)
SMILES O=C(C1=CC=C2C3=C(C(NC4=CC=CC(Cl)=C4)=NC2=C1)C=CN=C3)O[Na]
Molecular Weight (g/mol) 371.75
Formula Weight 371.75
Percent Purity 98.72%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.