missing translation for 'onlineSavingsMsg'
Learn More

Silmitasertib, MedChemExpress

Product Code. 16498108
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498108 10 mg 10mg
16480568 5 mg 5mg
16474027 50 mg 50mg
16447876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498108 Supplier Medchem Express Supplier No. HY50855
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.

Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.

Specifications

Chemical Name or Material Silmitasertib
CAS 1009820-21-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H12ClN3O2
Quantity 10 mg
Synonym CX-4945
Solubility Information DMSO : ≥ 35 mg/mL (100.07 mM) ∣0.1 M NaOH : 33.33 mg/mL (95.29 mM; ultrasonic and adjust pH to 9 with NaOH)
SMILES O=C(C1=CC=C2C3=C(C(NC4=CC=CC(Cl)=C4)=NC2=C1)C=CN=C3)O
Molecular Weight (g/mol) 349.77
Formula Weight 349.77
Percent Purity 96.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.