missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicone Viscosity Standard, 30,000mPas, Bookfield™
Description
A convenient, reliable means of checking the calibration of most Brookfield viscometers. These fluids are calibrated at 25°C in 500mL bottles and are supplied with instructions and calibration certificate. These Newtonian oils are ideal for the calibration of Brookfield viscometers and each one is traceable to NIST standards. Silicone oils should not be used in the paint and some food industries. Similarly silicone fluid should not be used for the calibration of glass viscometers due to coating of the viscometer walls.
- Accuracy: ±1%
- Excellent temperature stability
- Recommended for use with Brookfield and most other rotational viscometers
- Economical
Specifications
Specifications
| Packaging | Bottle |
| Viscosity | 30000mPas |
| Physical Form | Liquid |
| Quantity | 500 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?