Learn More
Welch Ilmvac™ Silicone Vacuum Hose
Flexible and resilient silicone hose suitable for vacuum pump use. Welch Ilmvac™ Silicone Vacuum Hose is durable and has an excellent lifespan.
Brand: Welch Ilmvac™ 828374
This item is not returnable.
View return policy
Description
- Suitable for vacuum pump use
- 12mm O.D.
- 6mm I.D.
- 3mm wall thickness