missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Silicone Tubing Pressure-sided

Product Code. 16438213 Shop All Sartorius Products
Click to view available options
Overall Length (English):
33 ft.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16438213

Brand: Sartorius 1ZAS0020

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Silicone Tubing Pressure-sided is connected to the Microsart™ E.jet Transfer Pump on the pressure side.

  • Microbiology Accessory
  • Required length depends on the distance between the transfer pump and the spout
TRUSTED_SUSTAINABILITY

Specifications

Material Silicone
Overall Length (English) 33 ft.
Overall Length (Metric) 10 m
Color Transparent
Product Type Pressure Tubing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.