missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Silicone Tubing

Product Code. 10361582 Shop All Heidolph Instruments Products
Change view
Click to view available options
Tubing Size:
0.5
4.8
6.4
Thickness (Metric):
1.6 mm
2.5 mm
Unit Size:
1m
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Thickness (Metric) unitSize
10361582 4.8 2.5 mm 1m
10721103 0.5 1.6 mm 1m
10660303 6.4 1.6 mm 1m
10585112 6.4 2.5 mm 1m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10361582 Supplier Heidolph™ Supplier No. 5253002700
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Platinum-coated silicone tubing for use in pharmaceuticals and biology. Heidolph™ Silicone Tubing is very smooth, biocompatible and inert.

  • Extremely smooth interior prevents bacterial growth.
  • Biocompatible, minimal adsorption and absorption.
  • Good flow properties.
  • High temperature stability.
  • Absolutely inert, softener-free.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Material Borosilicate Glass
Thickness (Metric) 2.5 mm
Sterility Non-sterile
Tubing Size 4.8
Maximum Pressure 0.8/1.8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.