Learn More
Lenz Laborglasintrumente Silicone Sealing Rings
Description
- For screw caps with bore
- Material: silicone rubber (VMQ)
- With bonded PTFE face
Specifications
Specifications
| Bore Size | 32 mm |
| Material | Silicone |