Learn More
BINDER™ Door Gasket
Shop All BINDER Products
Click to view available options
Material:
FKM Fluorocarbon
Unit Size:
Each
Description
- Manufactured from FKM fluorocarbon
- Temperature-resistant up to 200°C
Specifications
Specifications
| Description | Door gasket - made of FKM, silicone-free |
| For Use With (Equipment) | BINDER Chambers ED 400 |
| Material | FKM Fluorocarbon |
| Type | Door Gasket |