Learn More
BINDER™ Door Gasket
Product Code. 16404179 Shop All BINDER Products
Click to view available options
Material:
FKM Fluorocarbon
Unit Size:
Each
Product Code. 16404179
Brand: BINDER™ 80121310
Only null left
Add to basket
Add to basket
Material: FKM Fluorocarbon; For Use With: FED 400
  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications
Description Door gasket - made of FKM, silicone-free
For Use With (Equipment) BINDER Chambers FED 400
Material FKM Fluorocarbon
Type Door Gasket