Learn More
BINDER™ Door Gasket
Product Code. 16606483 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers VD/VDL 115
BINDER Chambers VD/VDL 23
BINDER Chambers VD/VDL 56
Unit Size:
Each
Product Code. 16606483
Brand: BINDER™ 86120009
Only null left
Add to basket
Add to basket
Door gasket - made of FKM, silicone-free
  • Perfectly matched, durable gasket
Specifications
Description Door gasket - made of FKM, silicone-free
For Use With (Equipment) BINDER Chambers VD/VDL 115
Material FKM Fluorocarbon
Type Door Gasket