missing translation for 'onlineSavingsMsg'
Learn More

Silicon Standard, 1000mg/L in H2O/0.24 F (Assurance Grade), SPEX CertiPrep™

Product Code. 10293952 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
30 mL
250 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
Unit Size:
250mL
30mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10293952 250 mL Poly Bottle 250mL
16301256 30 mL Triple-leached LDPE bottle 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10293952 Supplier SPEX Certiprep Supplier No. PLSI92T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Silicon Standard
  • Made with ASTM Type I Water
  • 1,000 μg/mL concentration
  • 250 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Chemical Identifiers

CAS 16919-19-0 , 7732-18-5
Molecular Formula F6H8N2Si
Molecular Weight (g/mol) 178.153
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
PubChem CID 28145
IUPAC Name diazanium;hexafluorosilicon(2-)
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F

Specifications

Packaging Poly Bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Technique AA, ICP
Type Certified Reference Material
Quantity 250 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.H2O/tr. HF    1000μg/mL    PLSI9-2Y, -2T, -2X    10,000    PLSI9-3Y, -3XH2O    1000μg/mL    PLSI9A-2X    10,000    PLSI9A-3X
Chemical Name or Material Silicon Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.