missing translation for 'onlineSavingsMsg'
Learn More

Silicon sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.999% (metals basis), Thermo Scientific™

Product Code. 15424825
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15424825

Brand: Thermo Scientific Alfa Aesar 040893.KS

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-21-3
Quantity 1 Ea.
Odor Odorless
Molecular Formula Si
MDL Number MFCD00085311
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
Solubility Information Insoluble in water.
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES [Si]
IUPAC Name silicon
Molecular Weight (g/mol) 28.09
PubChem CID 5461123
ChEBI CHEBI:27573
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Silicon sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.