Learn More
Thermo Scientific Alfa Aesar Silicon sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis)
Brand:  Thermo Scientific Alfa Aesar 040892.KS
Silicon sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-21-3 Weight : 0.01000kg
Product Code. 15414825