missing translation for 'onlineSavingsMsg'
Learn More

Silicon rod, 5.08cm (2.0 in.) dia., 99.999% (metals basis), Thermo Scientific™

Product Code. 11353478
Click to view available options
Quantity:
25 mm
100 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Quantity 25 mm
Percent Purity 99.999%
Odor Odorless
Weight ≈47.18g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Silicon rod, 5.08cm (2.0 in.) dia.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'