missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silicon plate, 9.5mm (0.37 in.) thick, 99.999% (metals basis)

Brand:  Alfa Aesar™ 039110.FI

Code : NEW

Additional Details : CAS Number : 7440-21-3 Weight : 0.01000kg

Quantity 50 x 50mm

 View more versions of this product

Product Code. 11460211

  • £226.60 / 50mm


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Specifications

Chemical Identifiers

Silicon plate
7440-21-3
9.5mm (0.37 in.) thick
MFCD00085311

Specifications

Silicon plate
50 x 50mm
7440-21-3
Plate
(metals basis)
≈14.32g/25x25mm
9.5mm (0.37 in.) thick
Odorless
99.999%
MFCD00085311
Videos
SDS