missing translation for 'onlineSavingsMsg'
Learn More

Silicon plate, 6.35mm (0.25in) thick, 99.999% (metals basis)

Product Code. 11363488
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
100mm
25mm
50mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363488 100 x 100 mm 100mm
11353488 25 x 25 mm 25mm
11470211 50 x 50 mm 50mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363488 Supplier Thermo Scientific Alfa Aesar Supplier No. 039112.GH
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-21-3 | Si | 28.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Quantity 100 x 100 mm
Odor Odorless
Weight ≈9.54g/25x25mm
Form Plate
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Silicon plate, 6.35mm (0.25 in.) thick

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.