missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)

Product Code. 11400051
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400051 100 g 100g
11410051 25 g 25g
11420051 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400051 Supplier Thermo Scientific Alfa Aesar Supplier No. 044781.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silicon(IV) oxide is used as sodium aluminate stabilizer. It is also used as SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. It is also acts as chain transfer agent in polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 60.09
Odor Odorless
Density 2.2 to 2.6 g/cm3 at 20°C
Chemical Name or Material Silicon(IV) oxide
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Silica, amorphous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.