missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11367007
Click to view available options
Quantity:
25 g
100 g
14808-60-7
O2Si
60.08
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
O=[Si]=O
14808-60-7
O2Si
60.08
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
O=[Si]=O

Silica is used in the manufacture of glass for windows, drinking glasses, beverage bottles, and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware, and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 2230°C
Melting Point 1710°C
Color Brown to White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.999% (metals basis)
Solubility Information Silica is poorly soluble in water. The solubility is higher for the amorphous form compared to the crystalline compound.
IUPAC Name dioxosilane
Formula Weight 60.09
Odor Odorless
Density 2.2-2.6g/mL
Chemical Name or Material Silicon(IV) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'