missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 99.0% (metals basis)

Product Code. 11423820
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423820 5 kg 5kg
11403820 250 g 250g
11413820 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423820 Supplier Thermo Scientific Alfa Aesar Supplier No. 089709.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Physical Form Fused Amorphous Powder
Quantity 5 kg
Assay Percent Range 99.8% (metals basis)
Solubility Information Insoluble in water.
IUPAC Name dioxosilane
Formula Weight 60.09
Density 2.2 to 2.6 g/mL
Chemical Name or Material Silicon(IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.