missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 50% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals

Catalog Number 11314778
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy
7631-86-9
O2Si
60.08
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
dioxosilane
O=[Si]=O
This item is not returnable. View return policy
7631-86-9
O2Si
60.08
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
dioxosilane
O=[Si]=O

S.A. 180m2/g

Silica is used primarily in the production of glass for windows, drinking glasses, beverage bottles, and many other uses. The majority of optical fibers for telecommunication are also made from silica. It is a primary raw material for many ceramics such as earthenware, stoneware, and porcelain. The main ingredient in sand casting for the manufacture of a large number of metallic components in engineering and other applications. Primarily as a flow agent in powdered foods, or to adsorb water in hygroscopic applications. It is the primary component of diatomaceous earth. Colloidal silica is also used as a wine, beer, and juice fining agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Particles in Liquid
Quantity 1 kg
Solubility Information Fully miscible in water. Soluble in alkalis.
Formula Weight 60.09
Odor Odorless
Density 1.40 g/cm3 at 20°C
Chemical Name or Material Silicon(IV) oxide, 50% in H2O

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'