missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 30% in Ethylene glycol, colloidal dispersion, Thermo Scientific Chemicals

Product Code. 11364748
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Product Code. Quantity unitSize
11364748 1 kg 1kg
11354748 5 kg 5kg
2 options
This item is not returnable. View return policy

Product Code. 11364748

Brand: Thermo Scientific Alfa Aesar 043091.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Physical Form Particles in Liquid
Quantity 1 kg
Linear Formula SiO2
Solubility Information Miscible in water. Partially miscible in alcohols,ketones. Immiscible in hydrocarbons.
Formula Weight 60.09
Concentration or Composition (by Analyte or Components) 30% in Ethylene glycol
Packaging Plastic bottle
Specific Gravity 1.3
Density 1.3
Chemical Name or Material Silicon(IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.