missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) iodide, 99.999% (metals basis)

Product Code. p-7059183
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11329638

Brand: Thermo Scientific Alfa Aesar 044141.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13465-84-4 | I4Si | 535.70 g/mol

Silicon(IV) iodide is used as a precursor to prepare silicon amides. It finds application in the manufacture and etching of silicon in microelectronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-84-4
Molecular Formula I4Si
Molecular Weight (g/mol) 535.70
MDL Number MFCD00054041
InChI Key CFTHARXEQHJSEH-UHFFFAOYSA-N
Synonym silicon tetraiodide, silane, tetraiodo, silicon iv iodide, periodosilane, tetraiodosilicon, acmc-1bu37, silicon tetraiodide trace metals basis, silicon iv iodide trace metals basis 1g
PubChem CID 83498
IUPAC Name tetraiodosilane
SMILES I[Si](I)(I)I

Specifications

Boiling Point 290°C
Melting Point 120°C
Color White
Physical Form Lump
Quantity 5 g
Linear Formula SiI4
UN Number UN2923
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 535.7
Percent Purity 99.999%
Sensitivity Air, light, and moisture sensitive
Density 4.198 g/mL
Assay (metals basis)
Chemical Name or Material Silicon (IV) iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.