missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) iodide, 99.999% (metals basis)

Product Code. 11339638
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339638 100 g 100g
11329638 5 g 5g
11319638 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339638 Supplier Thermo Scientific Alfa Aesar Supplier No. 044141.22
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13465-84-4 | I4Si | 535.70 g/mol

Silicon(IV) iodide is used as a precursor to prepare silicon amides. It finds application in the manufacture and etching of silicon in microelectronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-84-4
Molecular Formula I4Si
Molecular Weight (g/mol) 535.70
MDL Number MFCD00054041
InChI Key CFTHARXEQHJSEH-UHFFFAOYSA-N
Synonym silicon tetraiodide, silane, tetraiodo, silicon iv iodide, periodosilane, tetraiodosilicon, acmc-1bu37, silicon tetraiodide trace metals basis, silicon iv iodide trace metals basis 1g
PubChem CID 83498
IUPAC Name tetraiodosilane
SMILES I[Si](I)(I)I

Specifications

Boiling Point 290°C
Melting Point 120°C
Color White
Physical Form Lump
Quantity 100 g
Linear Formula SiI4
UN Number UN2923
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 535.7
Percent Purity 99.999%
Sensitivity Air, light, and moisture sensitive
Density 4.198 g/mL
Assay (metals basis)
Chemical Name or Material Silicon (IV) iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.