missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) chloride, 99%

Product Code. 11374799
Change view
Click to view available options
Quantity:
10 mL
100 mL
1 L
3 x 1 L
Unit Size:
1 L
100mL
10mL
3 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374799 1 L 1 L
11384799 10 mL 10mL
11364799 100 mL 100mL
11394799 3 x 1 L 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11374799 Supplier Thermo Scientific Alfa Aesar Supplier No. 069182.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10026-04-7 | Cl4Si | 169.89 g/mol

Silicon(IV) chloride is used as an intermediate in the manufacture of polysilicon. It is also used in the semiconductors industry, photovoltaic cells and in the production of optical fibers. Its derivatives like trichlorosilane and silane are used in the Siemens process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-04-7
Molecular Formula Cl4Si
Molecular Weight (g/mol) 169.89
MDL Number MFCD00011229
InChI Key FDNAPBUWERUEDA-UHFFFAOYSA-N
Synonym silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4
PubChem CID 24816
IUPAC Name tetrachlorosilane
SMILES Cl[Si](Cl)(Cl)Cl

Specifications

Boiling Point 57.6°C
Melting Point °C to 70°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Assay Percent Range 99% (metals basis)
Solubility Information Miscible with benzene,toluene,chloroform,petroleum ether,carbon tetrachloride,ether and hydrochloric acid.
Formula Weight 169.9
Odor Acrid
Sensitivity Moisture sensitive
Appearance Colourless liquid
Density 1.483
Chemical Name or Material Silicon(IV) chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silicon(IV) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: Silicon tetrachloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.