missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) bromide

Product Code. 11381559
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381559 10 g 10g
11391559 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381559 Supplier Thermo Scientific Alfa Aesar Supplier No. 014029.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-66-4 | Br4Si | 347.701 g/mol

Silicon(IV) bromide is used to grow thin films of silicon by the chemical vapor deposition method. It is also used for ceramics, sealants and the production of many cutting tools.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-66-4
Molecular Formula Br4Si
Molecular Weight (g/mol) 347.701
MDL Number MFCD00049530
InChI Key AIFMYMZGQVTROK-UHFFFAOYSA-N
Synonym silicon iv bromide, silicon tetrabromide, silane, tetrabromo, sibr4, unii-8gac9ywl42, 8gac9ywl42, silicon bromide, acmc-20ajqj
PubChem CID 82247
IUPAC Name tetrabromosilane
SMILES [Si](Br)(Br)(Br)Br

Specifications

Boiling Point 154°C
Melting Point 5°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 10 g
UN Number UN3264
Merck Index 14,8499
Formula Weight 347.72
Odor Stench
Sensitivity Moisture sensitive
Density 2.800 g/mL
Chemical Name or Material Silicon(IV) bromide
Show More Show Less
compliance-icons
Product Identifier
  • Silicon(IV) bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Silicon(IV) bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.