missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) acetate

Product Code. 11347008
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347008 50 g 50g
11357008 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347008 Supplier Thermo Scientific Alfa Aesar Supplier No. 014703.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 562-90-3 | C8H12O8Si | 264.26 g/mol

Silicon tetraacetate is used in the preparation of silicon dioxide thin films by a direct photochemical vapor deposition method. It serves as a precursor to prepare silicon complexes with monofunctional bidentate Schiff bases. It is also used as an alternative to silicon hydride and alkoxide for low-temperature silicon dioxide production. Further, it reacts with ethanol in the absence of water to get silica gel and ethyl acetate. In addition, it is employed as a sol-gel precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 562-90-3
Molecular Formula C8H12O8Si
Molecular Weight (g/mol) 264.26
MDL Number MFCD00026184
InChI Key YZVRVDPMGYFCGL-UHFFFAOYSA-N
Synonym silicon tetraacetate, tetraacetoxysilane, tetraacetyl orthosilicate, unii-vz7lp47epp, vz7lp47epp, silicon iv acetate, anhydrous, acetic acid, 1,1',1,1'-tetraanhydride with silicic acid h4sio4, triacetoxysilyl acetate, silanetetrol, tetraacetate
PubChem CID 68419
IUPAC Name triacetyloxysilyl acetate
SMILES CC(=O)O[Si](OC(C)=O)(OC(C)=O)OC(C)=O

Specifications

Boiling Point 160°C to 170°C (decomposition)
Melting Point 110°C
Physical Form Crystalline
Quantity 50 g
UN Number UN3261
Merck Index 14,8498
Solubility Information Soluble in acetone and benzene.
Formula Weight 264.27
Sensitivity Moisture sensitive
Chemical Name or Material Silicon(IV) acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.