missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon(II) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
A major application of silicon oxide is production of portland cement. It is used in food and pharmaceutical applications and in semiconductor industry. It is the major precursor for preparing glass and silicon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1880°C |
Melting Point | >1702°C |
Color | Black |
Physical Form | Lump |
Quantity | 25 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 44.09 |
Odor | Odorless |
Density | 2.13 g/cm3 at 20°C |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'