missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon diOxide, 99%, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 500 g |
| Linear Formula | SiO2 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 60.08g/mol |
| Percent Purity | 99% |
| Chemical Name or Material | Silicon dioxide |
Safety and Handling
Product Identifier
- Silicon dioxide
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-[In case of inadequate ventilation] wear respiratory protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Quartz
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?