missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide powder, fine, 320 grit

Product Code. 11447947
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447947 2500 g 2500g
11437947 500 g 500g
11457947 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447947 Supplier Thermo Scientific Alfa Aesar Supplier No. A16601.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 409-21-2 | CSi | 40.10 g/mol

Silicon carbide is used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon carbide is used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.

Solubility
Soluble in molten sodium hydroxide, potassium hydroxide and in molten iron.

Notes
Keep it away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
IUPAC Name methanidylidynesilanylium
SMILES [C-]#[Si+]

Specifications

Quantity 2500 g
Solubility Information Soluble in molten sodium hydroxide,potassium hydroxide and in molten iron.
Formula Weight 40.1
Odor Odorless
Density 3.16
Chemical Name or Material Silicon carbide powder, fine, 320 grit

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.